Gene Transcripts

Gene Symbol
TTK
Entrez Gene ID
7272
Gene Name
TTK protein kinase
Gene Aliases
CT96, ESK, MPH1, MPS1, MPS1L1, PYT
Chromosome Location
Chr. 6: 80004147 - 80043577 on Build GRCh38
UniGene ID
Hs.169840
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166691.1 NP_001160163.1 8 939
NM_003318.4 NP_003309.2 8 927
XM_011536099.2 XP_011534401.1 8 901
XM_011536100.2 XP_011534402.1 8 927
XM_017011242.1 XP_016866731.1 9 1456
GenBank mRNA AK095599.1 - 8 1399
AK292460.1 - 8 902
AK315696.1 - 8 844
AX775733.1 - 8 899
BC000633.2 - 8 890 3343925
BC032858.2 - 8 892 5270012

View Full Product Details