Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004870.3 | NP_004861.2 | 4 | 543 | |
NR_024603.1 | 4 | 538 | |||
XM_006721597.2 | XP_006721660.1 | 4 | 342 | ||
XM_006721598.2 | XP_006721661.1 | 4 | 342 | ||
XM_006721599.2 | XP_006721662.1 | 4 | 342 | ||
XM_011524081.2 | XP_011522383.1 | 4 | 374 | ||
GenBank mRNA | AF038961.1 | - | 4 | 343 | |
AF059752.1 | - | 4 | 383 | ||
AF258568.1 | - | 1 | 248 | ||
AK055923.1 | - | 4 | 337 | ||
AK075299.1 | - | 4 | 342 | ||
AK225401.1 | - | 4 | 342 | ||
AK225432.1 | - | 4 | 338 | ||
AK297001.1 | - | 4 | 338 | ||
AK297691.1 | - | 4 | 338 | ||
AK300140.1 | - | 4 | 329 | ||
AK314527.1 | - | 4 | 338 | ||
BC001898.1 | - | 4 | 354 | 3536216 | |
DC421138.1 | - | 4 | 544 | ||
DQ499597.1 | - | 4 | 327 | ||
KU178478.1 | - | 4 | 327 | ||
KU178479.1 | - | 4 | 363 |