Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26463835-26464336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530491.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2935537 c.1466C>T p.P489L Chr.22: 26464203-26464203 on Build GRCh38
4691091 c.1539C>T p.S513S Chr.22: 26464130-26464130 on Build GRCh38
5651995 c.1519G>C p.E507Q Chr.22: 26464111-26464111 on Build GRCh38
4103179 c.1720C>T p.L574L Chr.22: 26463949-26463949 on Build GRCh38
6681973 c.1525G>A p.G509R Chr.22: 26464144-26464144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772234990 Chr.22: 26464176-26464176 on Build GRCh38
rs868526867 Chr.22: 26464241-26464241 on Build GRCh38
rs367888424 Chr.22: 26463997-26463997 on Build GRCh38
rs549290787 Chr.22: 26464219-26464219 on Build GRCh38
rs112936259 Chr.22: 26463888-26463888 on Build GRCh38
rs372833027 Chr.22: 26464084-26464084 on Build GRCh38
rs186173240 Chr.22: 26464270-26464270 on Build GRCh38
rs763117738 Chr.22: 26464039-26464039 on Build GRCh38
rs772205948 Chr.22: 26464247-26464247 on Build GRCh38
rs886490917 Chr.22: 26463856-26463856 on Build GRCh38
rs367810755 Chr.22: 26464097-26464097 on Build GRCh38
rs965241113 Chr.22: 26464222-26464222 on Build GRCh38
rs958272499 Chr.22: 26463972-26463972 on Build GRCh38
rs764305334 Chr.22: 26464044-26464044 on Build GRCh38
rs2014410 Chr.22: 26464303-26464303 on Build GRCh38
rs983056340 Chr.22: 26464254-26464254 on Build GRCh38
rs757988379 Chr.22: 26464150-26464150 on Build GRCh38
rs143902143 Chr.22: 26463951-26463951 on Build GRCh38
rs777570870 Chr.22: 26464151-26464151 on Build GRCh38
rs758568345 Chr.22: 26463964-26463964 on Build GRCh38
rs748736760 Chr.22: 26464005-26464005 on Build GRCh38
rs757752164 Chr.22: 26463901-26463901 on Build GRCh38
rs943099617 Chr.22: 26464003-26464003 on Build GRCh38
rs5752330 Chr.22: 26463976-26463976 on Build GRCh38
rs540157835 Chr.22: 26463971-26463971 on Build GRCh38
rs747323111 Chr.22: 26463900-26463900 on Build GRCh38
rs765633123 Chr.22: 26464297-26464297 on Build GRCh38
rs182592572 Chr.22: 26463988-26463988 on Build GRCh38
rs774813995 Chr.22: 26463927-26463927 on Build GRCh38
rs769325136 Chr.22: 26463923-26463923 on Build GRCh38
rs773570398 Chr.22: 26464180-26464180 on Build GRCh38
rs748133574 Chr.22: 26464016-26464016 on Build GRCh38
rs764860798 Chr.22: 26464216-26464216 on Build GRCh38
rs751013277 Chr.22: 26464305-26464305 on Build GRCh38
rs756512740 Chr.22: 26464076-26464076 on Build GRCh38
rs773303744 Chr.22: 26464252-26464252 on Build GRCh38
rs774728501 Chr.22: 26464166-26464166 on Build GRCh38
rs1021234084 Chr.22: 26464164-26464164 on Build GRCh38
rs755055492 Chr.22: 26463872-26463872 on Build GRCh38
rs745687268 Chr.22: 26464163-26464163 on Build GRCh38
rs139039617 Chr.22: 26464212-26464212 on Build GRCh38
rs1037852081 Chr.22: 26463874-26463874 on Build GRCh38
rs772069598 Chr.22: 26464024-26464024 on Build GRCh38
rs763704933 Chr.22: 26464228-26464228 on Build GRCh38
rs751051493 Chr.22: 26464231-26464231 on Build GRCh38
rs781082499 Chr.22: 26464158-26464158 on Build GRCh38
rs759632995 Chr.22: 26463938-26463938 on Build GRCh38
rs778431397 Chr.22: 26463896-26463896 on Build GRCh38
rs534067573 Chr.22: 26464127-26464127 on Build GRCh38
rs768931718 Chr.22: 26464165-26464165 on Build GRCh38
rs772846606 Chr.22: 26464026-26464026 on Build GRCh38
rs767256800 Chr.22: 26463935-26463935 on Build GRCh38
rs770823084 Chr.22: 26464152-26464152 on Build GRCh38
rs766903925 Chr.22: 26464188-26464188 on Build GRCh38
rs781455905 Chr.22: 26463980-26463980 on Build GRCh38
rs765322078 Chr.22: 26464033-26464033 on Build GRCh38
rs767548244 Chr.22: 26464314-26464314 on Build GRCh38
rs564289679 Chr.22: 26463880-26463880 on Build GRCh38
rs942076085 Chr.22: 26464173-26464173 on Build GRCh38
rs760527580 Chr.22: 26464028-26464028 on Build GRCh38
rs150216540 Chr.22: 26464095-26464095 on Build GRCh38
rs375587915 Chr.22: 26464260-26464260 on Build GRCh38
rs763311479 Chr.22: 26464110-26464110 on Build GRCh38
rs532996881 Chr.22: 26463883-26463883 on Build GRCh38
rs988050148 Chr.22: 26464178-26464178 on Build GRCh38
rs767852159 Chr.22: 26464126-26464126 on Build GRCh38
rs768513194 Chr.22: 26463932-26463932 on Build GRCh38
rs561308139 Chr.22: 26463936-26463936 on Build GRCh38
rs774532334 Chr.22: 26464119-26464119 on Build GRCh38
rs145674158 Chr.22: 26463990-26463990 on Build GRCh38
rs532246881 Chr.22: 26463993-26463993 on Build GRCh38
rs141008676 Chr.22: 26464094-26464094 on Build GRCh38
rs527653764 Chr.22: 26463950-26463950 on Build GRCh38
rs148134252 Chr.22: 26464087-26464087 on Build GRCh38
rs745543124 Chr.22: 26463920-26463920 on Build GRCh38
rs780889236 Chr.22: 26463919-26463919 on Build GRCh38
rs142958812 Chr.22: 26464233-26464233 on Build GRCh38
rs1038656474 Chr.22: 26463860-26463860 on Build GRCh38
rs770822747 Chr.22: 26464103-26464103 on Build GRCh38
rs752092814 Chr.22: 26464148-26464148 on Build GRCh38
rs893836600 Chr.22: 26464125-26464125 on Build GRCh38
rs756030696 Chr.22: 26464236-26464236 on Build GRCh38
rs1046054798 Chr.22: 26464308-26464308 on Build GRCh38
rs147435410 Chr.22: 26464234-26464234 on Build GRCh38
rs199740072 Chr.22: 26463989-26463989 on Build GRCh38
rs768468362 Chr.22: 26464277-26464279 on Build GRCh38
rs775916508 Chr.22: 26464034-26464034 on Build GRCh38
rs11545917 Chr.22: 26464143-26464143 on Build GRCh38
rs373176483 Chr.22: 26463907-26463907 on Build GRCh38
rs116241923 Chr.22: 26464015-26464015 on Build GRCh38
rs957749322 Chr.22: 26464269-26464269 on Build GRCh38
rs754462097 Chr.22: 26464091-26464091 on Build GRCh38
rs886298215 Chr.22: 26463914-26463914 on Build GRCh38
rs761197874 Chr.22: 26464187-26464187 on Build GRCh38
rs200667267 Chr.22: 26464049-26464049 on Build GRCh38
rs752086428 Chr.22: 26464046-26464046 on Build GRCh38
rs752847714 Chr.22: 26463881-26463881 on Build GRCh38
rs778910458 Chr.22: 26463873-26463873 on Build GRCh38
rs778544549 Chr.22: 26464142-26464142 on Build GRCh38
rs774331295 Chr.22: 26463934-26463934 on Build GRCh38
rs748136926 Chr.22: 26464274-26464274 on Build GRCh38
rs754224646 Chr.22: 26464137-26464137 on Build GRCh38
rs769017976 Chr.22: 26464116-26464116 on Build GRCh38
rs756839100 Chr.22: 26464310-26464310 on Build GRCh38
rs751938775 Chr.22: 26463994-26463994 on Build GRCh38
rs768100817 Chr.22: 26464073-26464073 on Build GRCh38
rs756215705 Chr.22: 26464175-26464175 on Build GRCh38
rs1034179775 Chr.22: 26464120-26464120 on Build GRCh38
rs754066787 Chr.22: 26464245-26464245 on Build GRCh38
rs143747386 Chr.22: 26464059-26464059 on Build GRCh38
rs372027593 Chr.22: 26464235-26464235 on Build GRCh38
rs757812079 Chr.22: 26464227-26464227 on Build GRCh38
rs748839497 Chr.22: 26463898-26463898 on Build GRCh38
rs775820827 Chr.22: 26464106-26464106 on Build GRCh38
rs202104505 Chr.22: 26464009-26464009 on Build GRCh38
rs766781092 Chr.22: 26464128-26464128 on Build GRCh38
rs778346173 Chr.22: 26464092-26464092 on Build GRCh38
rs761956374 Chr.22: 26464121-26464121 on Build GRCh38
rs774979651 Chr.22: 26464300-26464300 on Build GRCh38
rs140234219 Chr.22: 26463930-26463930 on Build GRCh38
rs762613783 Chr.22: 26464301-26464301 on Build GRCh38
rs759909785 Chr.22: 26464213-26464213 on Build GRCh38
rs994746146 Chr.22: 26463885-26463885 on Build GRCh38
rs778998363 Chr.22: 26464243-26464243 on Build GRCh38
rs750685060 Chr.22: 26464074-26464074 on Build GRCh38

View Full Product Details