Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15456223-15456750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
UniGene
Hs.146735
Gene Chromosome Location
Chr.3: 15450133-15521751 on Build GRCh38
Transcripts
NM_005677.3 NM_080538.2 NM_080539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5658436 c.1015A>G p.K339E Chr.3: 15456489-15456489 on Build GRCh38
7213398 c.988C>T p.R330C Chr.3: 15456516-15456516 on Build GRCh38
3589022 c.1006C>T p.L336L Chr.3: 15456498-15456498 on Build GRCh38
3731720 c.1008C>T p.A336A Chr.3: 15456526-15456526 on Build GRCh38
7260590 c.1044+8delC p.? Chr.3: 15456452-15456452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762939356 Chr.3: 15456418-15456418 on Build GRCh38
rs577208764 Chr.3: 15456274-15456275 on Build GRCh38
rs1035415490 Chr.3: 15456626-15456626 on Build GRCh38
rs959716120 Chr.3: 15456313-15456313 on Build GRCh38
rs932015384 Chr.3: 15456408-15456408 on Build GRCh38
rs138818971 Chr.3: 15456516-15456516 on Build GRCh38
rs924593823 Chr.3: 15456699-15456699 on Build GRCh38
rs372064982 Chr.3: 15456462-15456462 on Build GRCh38
rs139329020 Chr.3: 15456256-15456257 on Build GRCh38
rs149532541 Chr.3: 15456503-15456503 on Build GRCh38
rs375225755 Chr.3: 15456515-15456515 on Build GRCh38
rs564085701 Chr.3: 15456658-15456658 on Build GRCh38
rs113708721 Chr.3: 15456534-15456534 on Build GRCh38
rs868541965 Chr.3: 15456500-15456500 on Build GRCh38
rs79557839 Chr.3: 15456332-15456332 on Build GRCh38
rs574707779 Chr.3: 15456326-15456326 on Build GRCh38
rs1023169531 Chr.3: 15456655-15456656 on Build GRCh38
rs1011814904 Chr.3: 15456550-15456550 on Build GRCh38
rs1021423005 Chr.3: 15456563-15456563 on Build GRCh38
rs77027767 Chr.3: 15456406-15456406 on Build GRCh38
rs1038736084 Chr.3: 15456365-15456365 on Build GRCh38
rs754194997 Chr.3: 15456476-15456476 on Build GRCh38
rs987318581 Chr.3: 15456725-15456725 on Build GRCh38
rs145130047 Chr.3: 15456681-15456681 on Build GRCh38
rs375072260 Chr.3: 15456446-15456446 on Build GRCh38
rs533487711 Chr.3: 15456665-15456665 on Build GRCh38
rs757815714 Chr.3: 15456495-15456495 on Build GRCh38
rs375833598 Chr.3: 15456288-15456289 on Build GRCh38
rs760303798 Chr.3: 15456467-15456467 on Build GRCh38
rs779583406 Chr.3: 15456497-15456497 on Build GRCh38
rs111725562 Chr.3: 15456274-15456274 on Build GRCh38
rs201036462 Chr.3: 15456564-15456564 on Build GRCh38
rs576206818 Chr.3: 15456383-15456383 on Build GRCh38
rs372534357 Chr.3: 15456585-15456585 on Build GRCh38
rs762298520 Chr.3: 15456586-15456586 on Build GRCh38
rs983160963 Chr.3: 15456682-15456682 on Build GRCh38
rs1035277761 Chr.3: 15456280-15456280 on Build GRCh38
rs369551954 Chr.3: 15456567-15456567 on Build GRCh38
rs750978700 Chr.3: 15456616-15456616 on Build GRCh38
rs956240344 Chr.3: 15456720-15456720 on Build GRCh38
rs770819352 Chr.3: 15456529-15456529 on Build GRCh38
rs372296111 Chr.3: 15456345-15456345 on Build GRCh38
rs200349012 Chr.3: 15456621-15456621 on Build GRCh38
rs969812260 Chr.3: 15456265-15456265 on Build GRCh38
rs750489224 Chr.3: 15456498-15456498 on Build GRCh38
rs770907653 Chr.3: 15456424-15456424 on Build GRCh38
rs201227079 Chr.3: 15456456-15456456 on Build GRCh38
rs749107627 Chr.3: 15456517-15456517 on Build GRCh38
rs760343679 Chr.3: 15456565-15456565 on Build GRCh38
rs752410020 Chr.3: 15456465-15456465 on Build GRCh38
rs372275872 Chr.3: 15456414-15456414 on Build GRCh38
rs758991725 Chr.3: 15456547-15456547 on Build GRCh38
rs772934171 Chr.3: 15456411-15456411 on Build GRCh38
rs765770364 Chr.3: 15456597-15456597 on Build GRCh38
rs546057149 Chr.3: 15456335-15456335 on Build GRCh38
rs771559323 Chr.3: 15456552-15456552 on Build GRCh38
rs764516134 Chr.3: 15456472-15456472 on Build GRCh38
rs766472634 Chr.3: 15456628-15456628 on Build GRCh38
rs759028362 Chr.3: 15456627-15456627 on Build GRCh38
rs368069106 Chr.3: 15456327-15456327 on Build GRCh38
rs758554049 Chr.3: 15456508-15456508 on Build GRCh38
rs1015584162 Chr.3: 15456684-15456684 on Build GRCh38
rs184944796 Chr.3: 15456510-15456510 on Build GRCh38
rs867514391 Chr.3: 15456632-15456632 on Build GRCh38
rs886108080 Chr.3: 15456519-15456519 on Build GRCh38
rs774279832 Chr.3: 15456535-15456535 on Build GRCh38
rs562575822 Chr.3: 15456354-15456354 on Build GRCh38
rs113488563 Chr.3: 15456556-15456556 on Build GRCh38

View Full Product Details