Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6012346-6012849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

SNP Detail

SNP ID Chromosome Location
rs572213760 Chr.10: 6012670-6012670 on Build GRCh38
rs753838977 Chr.10: 6012773-6012773 on Build GRCh38
rs886047080 Chr.10: 6012769-6012769 on Build GRCh38
rs903627253 Chr.10: 6012657-6012657 on Build GRCh38
rs866767255 Chr.10: 6012647-6012647 on Build GRCh38
rs972136274 Chr.10: 6012780-6012780 on Build GRCh38
rs553112485 Chr.10: 6012401-6012401 on Build GRCh38
rs12722600 Chr.10: 6012802-6012802 on Build GRCh38
rs943884205 Chr.10: 6012636-6012636 on Build GRCh38
rs181357994 Chr.10: 6012411-6012411 on Build GRCh38
rs12722601 Chr.10: 6012428-6012428 on Build GRCh38
rs990982402 Chr.10: 6012436-6012436 on Build GRCh38
rs886047081 Chr.10: 6012779-6012779 on Build GRCh38
rs1023976376 Chr.10: 6012715-6012715 on Build GRCh38
rs984852514 Chr.10: 6012402-6012402 on Build GRCh38
rs1043477456 Chr.10: 6012634-6012634 on Build GRCh38
rs559720581 Chr.10: 6012633-6012633 on Build GRCh38
rs768574986 Chr.10: 6012419-6012419 on Build GRCh38
rs12722715 Chr.10: 6012549-6012549 on Build GRCh38
rs886047078 Chr.10: 6012458-6012458 on Build GRCh38
rs910851011 Chr.10: 6012542-6012542 on Build GRCh38
rs188613092 Chr.10: 6012394-6012394 on Build GRCh38
rs575663406 Chr.10: 6012822-6012822 on Build GRCh38
rs919824754 Chr.10: 6012585-6012585 on Build GRCh38
rs1040799650 Chr.10: 6012637-6012637 on Build GRCh38
rs999339677 Chr.10: 6012662-6012662 on Build GRCh38
rs1012922758 Chr.10: 6012755-6012755 on Build GRCh38
rs931239603 Chr.10: 6012613-6012613 on Build GRCh38
rs970867521 Chr.10: 6012779-6012779 on Build GRCh38
rs970752915 Chr.10: 6012464-6012464 on Build GRCh38
rs886047079 Chr.10: 6012461-6012462 on Build GRCh38
rs1024227901 Chr.10: 6012446-6012446 on Build GRCh38
rs1013475322 Chr.10: 6012691-6012691 on Build GRCh38
rs751588150 Chr.10: 6012558-6012558 on Build GRCh38
rs1023612429 Chr.10: 6012389-6012389 on Build GRCh38
rs909326184 Chr.10: 6012410-6012410 on Build GRCh38
rs756213567 Chr.10: 6012567-6012567 on Build GRCh38
rs923460298 Chr.10: 6012479-6012479 on Build GRCh38
rs12722714 Chr.10: 6012765-6012765 on Build GRCh38
rs547047548 Chr.10: 6012811-6012811 on Build GRCh38
rs34686279 Chr.10: 6012489-6012490 on Build GRCh38
rs576607759 Chr.10: 6012384-6012384 on Build GRCh38
rs531582979 Chr.10: 6012723-6012723 on Build GRCh38
rs1050646874 Chr.10: 6012537-6012537 on Build GRCh38
rs933546058 Chr.10: 6012517-6012517 on Build GRCh38
rs749452297 Chr.10: 6012768-6012768 on Build GRCh38
rs185829590 Chr.10: 6012598-6012598 on Build GRCh38
rs958546994 Chr.10: 6012432-6012432 on Build GRCh38
rs972229492 Chr.10: 6012443-6012443 on Build GRCh38
rs542942584 Chr.10: 6012689-6012689 on Build GRCh38
rs535911457 Chr.10: 6012663-6012663 on Build GRCh38
rs574873827 Chr.10: 6012477-6012477 on Build GRCh38
rs982154177 Chr.10: 6012489-6012489 on Build GRCh38
rs12722713 Chr.10: 6012817-6012817 on Build GRCh38
rs1053883274 Chr.10: 6012694-6012694 on Build GRCh38
rs866534259 Chr.10: 6012509-6012509 on Build GRCh38
rs560898947 Chr.10: 6012706-6012706 on Build GRCh38
rs549892288 Chr.10: 6012749-6012749 on Build GRCh38
rs962415501 Chr.10: 6012769-6012769 on Build GRCh38
rs778662335 Chr.10: 6012826-6012826 on Build GRCh38
rs973840090 Chr.10: 6012551-6012551 on Build GRCh38
rs367718232 Chr.10: 6012667-6012667 on Build GRCh38

View Full Product Details