Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6024054-6024569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3439119 c.276A>G p.K92K Chr.10: 6024335-6024335 on Build GRCh38
4903633 c.312G>A p.R104R Chr.10: 6024299-6024299 on Build GRCh38
1190819 c.307delG p.E103fs*68 Chr.10: 6024304-6024304 on Build GRCh38
6129761 c.357G>T p.A119A Chr.10: 6024254-6024254 on Build GRCh38
3439120 c.273G>A p.T91T Chr.10: 6024338-6024338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56054476 Chr.10: 6024279-6024279 on Build GRCh38
rs753336611 Chr.10: 6024247-6024247 on Build GRCh38
rs775602474 Chr.10: 6024211-6024211 on Build GRCh38
rs993981763 Chr.10: 6024457-6024457 on Build GRCh38
rs773086693 Chr.10: 6024332-6024332 on Build GRCh38
rs879788089 Chr.10: 6024498-6024499 on Build GRCh38
rs41290331 Chr.10: 6024338-6024338 on Build GRCh38
rs139340259 Chr.10: 6024348-6024348 on Build GRCh38
rs1048697267 Chr.10: 6024462-6024462 on Build GRCh38
rs376596128 Chr.10: 6024269-6024269 on Build GRCh38
rs780542426 Chr.10: 6024303-6024303 on Build GRCh38
rs770226758 Chr.10: 6024299-6024299 on Build GRCh38
rs140144132 Chr.10: 6024122-6024122 on Build GRCh38
rs750718897 Chr.10: 6024378-6024378 on Build GRCh38
rs376988755 Chr.10: 6024400-6024400 on Build GRCh38
rs201605377 Chr.10: 6024224-6024224 on Build GRCh38
rs72650666 Chr.10: 6024339-6024339 on Build GRCh38
rs372822702 Chr.10: 6024205-6024205 on Build GRCh38
rs1039738418 Chr.10: 6024439-6024439 on Build GRCh38
rs761616711 Chr.10: 6024233-6024233 on Build GRCh38
rs12358961 Chr.10: 6024232-6024232 on Build GRCh38
rs929482812 Chr.10: 6024365-6024365 on Build GRCh38
rs12722576 Chr.10: 6024454-6024454 on Build GRCh38
rs761978980 Chr.10: 6024330-6024330 on Build GRCh38
rs774606930 Chr.10: 6024329-6024329 on Build GRCh38
rs200838485 Chr.10: 6024311-6024311 on Build GRCh38
rs540440085 Chr.10: 6024471-6024471 on Build GRCh38
rs906569134 Chr.10: 6024187-6024187 on Build GRCh38
rs373756302 Chr.10: 6024183-6024183 on Build GRCh38
rs368295922 Chr.10: 6024223-6024223 on Build GRCh38
rs752391972 Chr.10: 6024358-6024358 on Build GRCh38
rs12722691 Chr.10: 6024186-6024186 on Build GRCh38
rs111237616 Chr.10: 6024166-6024166 on Build GRCh38
rs906401859 Chr.10: 6024547-6024547 on Build GRCh38
rs373292483 Chr.10: 6024241-6024241 on Build GRCh38
rs919465384 Chr.10: 6024171-6024171 on Build GRCh38
rs76029930 Chr.10: 6024273-6024273 on Build GRCh38
rs748276100 Chr.10: 6024405-6024405 on Build GRCh38
rs753853235 Chr.10: 6024340-6024340 on Build GRCh38
rs764176803 Chr.10: 6024410-6024410 on Build GRCh38
rs781478336 Chr.10: 6024274-6024274 on Build GRCh38
rs1045194390 Chr.10: 6024376-6024376 on Build GRCh38
rs200048148 Chr.10: 6024369-6024369 on Build GRCh38
rs866230452 Chr.10: 6024239-6024239 on Build GRCh38
rs980863658 Chr.10: 6024104-6024104 on Build GRCh38
rs12722574 Chr.10: 6024499-6024499 on Build GRCh38
rs150335219 Chr.10: 6024461-6024461 on Build GRCh38
rs751026252 Chr.10: 6024526-6024526 on Build GRCh38
rs759392020 Chr.10: 6024380-6024380 on Build GRCh38
rs74162095 Chr.10: 6024264-6024264 on Build GRCh38
rs752342176 Chr.10: 6024263-6024263 on Build GRCh38
rs144887378 Chr.10: 6024474-6024474 on Build GRCh38
rs1038233467 Chr.10: 6024150-6024150 on Build GRCh38
rs12722575 Chr.10: 6024498-6024498 on Build GRCh38
rs201105599 Chr.10: 6024314-6024314 on Build GRCh38
rs36065822 Chr.10: 6024338-6024339 on Build GRCh38
rs935511423 Chr.10: 6024097-6024097 on Build GRCh38
rs767262856 Chr.10: 6024234-6024234 on Build GRCh38
rs150164762 Chr.10: 6024254-6024254 on Build GRCh38
rs188218084 Chr.10: 6024417-6024417 on Build GRCh38
rs780486971 Chr.10: 6024384-6024384 on Build GRCh38
rs753848613 Chr.10: 6024425-6024425 on Build GRCh38
rs1024769629 Chr.10: 6024525-6024525 on Build GRCh38
rs771893707 Chr.10: 6024331-6024331 on Build GRCh38
rs11256369 Chr.10: 6024237-6024237 on Build GRCh38
rs749326313 Chr.10: 6024306-6024306 on Build GRCh38
rs12722690 Chr.10: 6024401-6024401 on Build GRCh38
rs1052045289 Chr.10: 6024203-6024203 on Build GRCh38
rs74899942 Chr.10: 6024514-6024674 on Build GRCh38
rs760755087 Chr.10: 6024238-6024238 on Build GRCh38
rs560423405 Chr.10: 6024519-6024519 on Build GRCh38
rs771222644 Chr.10: 6024202-6024202 on Build GRCh38
rs777493737 Chr.10: 6024374-6024374 on Build GRCh38
rs1039385689 Chr.10: 6024235-6024235 on Build GRCh38
rs1031218613 Chr.10: 6024537-6024537 on Build GRCh38
rs886041037 Chr.10: 6024310-6024310 on Build GRCh38
rs138645280 Chr.10: 6024255-6024255 on Build GRCh38
rs759710102 Chr.10: 6024121-6024121 on Build GRCh38
rs767805837 Chr.10: 6024188-6024188 on Build GRCh38
rs919412627 Chr.10: 6024349-6024349 on Build GRCh38
rs376969086 Chr.10: 6024210-6024210 on Build GRCh38
rs777175752 Chr.10: 6024204-6024204 on Build GRCh38
rs888325992 Chr.10: 6024464-6024464 on Build GRCh38

View Full Product Details