Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63060367-63060876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

SNP Detail

SNP ID Chromosome Location
rs750327382 Chr.8: 63060484-63060484 on Build GRCh38
rs986943054 Chr.8: 63060422-63060422 on Build GRCh38
rs1017467249 Chr.8: 63060799-63060799 on Build GRCh38
rs760669854 Chr.8: 63060657-63060657 on Build GRCh38
rs114278576 Chr.8: 63060470-63060470 on Build GRCh38
rs942657286 Chr.8: 63060429-63060429 on Build GRCh38
rs1019993995 Chr.8: 63060403-63060403 on Build GRCh38
rs187670490 Chr.8: 63060643-63060643 on Build GRCh38
rs185619905 Chr.8: 63060392-63060392 on Build GRCh38
rs539696099 Chr.8: 63060533-63060533 on Build GRCh38
rs917954788 Chr.8: 63060787-63060787 on Build GRCh38
rs905729592 Chr.8: 63060516-63060516 on Build GRCh38
rs181703462 Chr.8: 63060451-63060451 on Build GRCh38
rs544006849 Chr.8: 63060747-63060747 on Build GRCh38
rs974318768 Chr.8: 63060684-63060684 on Build GRCh38
rs555931173 Chr.8: 63060705-63060705 on Build GRCh38
rs886063065 Chr.8: 63060674-63060674 on Build GRCh38
rs1045580235 Chr.8: 63060481-63060481 on Build GRCh38
rs942947278 Chr.8: 63060445-63060445 on Build GRCh38
rs1021256777 Chr.8: 63060460-63060460 on Build GRCh38
rs931584885 Chr.8: 63060443-63060443 on Build GRCh38
rs1052928954 Chr.8: 63060727-63060727 on Build GRCh38
rs964174615 Chr.8: 63060546-63060546 on Build GRCh38
rs553790242 Chr.8: 63060456-63060456 on Build GRCh38
rs886063064 Chr.8: 63060517-63060517 on Build GRCh38
rs915614216 Chr.8: 63060673-63060674 on Build GRCh38
rs565583075 Chr.8: 63060756-63060756 on Build GRCh38
rs1020095376 Chr.8: 63060521-63060521 on Build GRCh38
rs577569514 Chr.8: 63060807-63060807 on Build GRCh38
rs983067188 Chr.8: 63060635-63060635 on Build GRCh38
rs576595274 Chr.8: 63060698-63060698 on Build GRCh38
rs552068173 Chr.8: 63060425-63060425 on Build GRCh38
rs762823451 Chr.8: 63060518-63060518 on Build GRCh38
rs190384822 Chr.8: 63060397-63060397 on Build GRCh38
rs955514479 Chr.8: 63060421-63060421 on Build GRCh38
rs766356779 Chr.8: 63060745-63060745 on Build GRCh38
rs775461769 Chr.8: 63060483-63060483 on Build GRCh38
rs975806349 Chr.8: 63060548-63060548 on Build GRCh38
rs987328217 Chr.8: 63060398-63060398 on Build GRCh38
rs996983591 Chr.8: 63060822-63060822 on Build GRCh38
rs964595146 Chr.8: 63060662-63060662 on Build GRCh38
rs76851526 Chr.8: 63060675-63060675 on Build GRCh38
rs886063066 Chr.8: 63060800-63060800 on Build GRCh38
rs35504965 Chr.8: 63060674-63060676 on Build GRCh38
rs920146723 Chr.8: 63060442-63060442 on Build GRCh38
rs1001450282 Chr.8: 63060517-63060517 on Build GRCh38
rs62510092 Chr.8: 63060607-63060607 on Build GRCh38
rs538656516 Chr.8: 63060444-63060444 on Build GRCh38
rs919950325 Chr.8: 63060588-63060588 on Build GRCh38
rs938808381 Chr.8: 63060652-63060652 on Build GRCh38
rs570386141 Chr.8: 63060436-63060436 on Build GRCh38
rs975457415 Chr.8: 63060435-63060435 on Build GRCh38
rs886063063 Chr.8: 63060476-63060476 on Build GRCh38
rs10543235 Chr.8: 63060674-63060674 on Build GRCh38
rs554751476 Chr.8: 63060647-63060647 on Build GRCh38
rs1009875834 Chr.8: 63060560-63060560 on Build GRCh38
rs949838244 Chr.8: 63060476-63060476 on Build GRCh38
rs929970250 Chr.8: 63060455-63060455 on Build GRCh38
rs768651971 Chr.8: 63060674-63060677 on Build GRCh38
rs892762690 Chr.8: 63060524-63060524 on Build GRCh38
rs952941213 Chr.8: 63060629-63060629 on Build GRCh38
rs986537749 Chr.8: 63060653-63060653 on Build GRCh38
rs1012970423 Chr.8: 63060458-63060458 on Build GRCh38
rs1019962925 Chr.8: 63060640-63060640 on Build GRCh38
rs752352118 Chr.8: 63060446-63060446 on Build GRCh38

View Full Product Details