Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63085636-63086136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5033560 c.204+2T>G p.? Chr.8: 63085816-63085816 on Build GRCh38
6365848 c.191A>G p.D64G Chr.8: 63085831-63085831 on Build GRCh38
6211965 c.106C>T p.R36W Chr.8: 63085916-63085916 on Build GRCh38
5780215 c.132G>A p.P44P Chr.8: 63085890-63085890 on Build GRCh38
289982 c.147C>G p.D49E Chr.8: 63085875-63085875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397515523 Chr.8: 63085831-63085831 on Build GRCh38
rs369019692 Chr.8: 63085751-63085751 on Build GRCh38
rs144290434 Chr.8: 63085965-63085965 on Build GRCh38
rs140010311 Chr.8: 63085998-63085998 on Build GRCh38
rs191599269 Chr.8: 63085769-63085769 on Build GRCh38
rs770593544 Chr.8: 63085982-63085982 on Build GRCh38
rs773895915 Chr.8: 63085834-63085834 on Build GRCh38
rs928950844 Chr.8: 63086089-63086089 on Build GRCh38
rs1026397217 Chr.8: 63086050-63086050 on Build GRCh38
rs763889339 Chr.8: 63085896-63085896 on Build GRCh38
rs780199989 Chr.8: 63085927-63085927 on Build GRCh38
rs760608813 Chr.8: 63086001-63086001 on Build GRCh38
rs866884489 Chr.8: 63085886-63085886 on Build GRCh38
rs950779916 Chr.8: 63086055-63086055 on Build GRCh38
rs781750837 Chr.8: 63085950-63085950 on Build GRCh38
rs1057516391 Chr.8: 63086009-63086009 on Build GRCh38
rs557316937 Chr.8: 63085830-63085830 on Build GRCh38
rs372863215 Chr.8: 63086104-63086104 on Build GRCh38
rs770318137 Chr.8: 63085771-63085771 on Build GRCh38
rs773429266 Chr.8: 63085759-63085759 on Build GRCh38
rs771592192 Chr.8: 63085992-63085992 on Build GRCh38
rs562202164 Chr.8: 63085974-63085974 on Build GRCh38
rs199805621 Chr.8: 63085875-63085875 on Build GRCh38
rs927622193 Chr.8: 63086102-63086102 on Build GRCh38
rs761564209 Chr.8: 63086059-63086059 on Build GRCh38
rs748690712 Chr.8: 63085957-63085957 on Build GRCh38
rs987756613 Chr.8: 63086033-63086033 on Build GRCh38
rs758641202 Chr.8: 63085920-63085920 on Build GRCh38
rs1024826881 Chr.8: 63086066-63086066 on Build GRCh38
rs973874631 Chr.8: 63085820-63085820 on Build GRCh38
rs1030533790 Chr.8: 63085770-63085770 on Build GRCh38
rs904907875 Chr.8: 63085663-63085663 on Build GRCh38
rs573070621 Chr.8: 63085938-63085938 on Build GRCh38
rs949291316 Chr.8: 63085883-63085883 on Build GRCh38
rs773911591 Chr.8: 63085986-63085986 on Build GRCh38
rs555671816 Chr.8: 63085773-63085773 on Build GRCh38
rs924900839 Chr.8: 63085761-63085761 on Build GRCh38
rs913616783 Chr.8: 63086115-63086115 on Build GRCh38
rs397515522 Chr.8: 63085847-63085847 on Build GRCh38
rs1018032856 Chr.8: 63085791-63085791 on Build GRCh38
rs936373434 Chr.8: 63085943-63085943 on Build GRCh38
rs764083716 Chr.8: 63086039-63086039 on Build GRCh38
rs1008240677 Chr.8: 63086009-63086009 on Build GRCh38
rs982650476 Chr.8: 63085850-63085850 on Build GRCh38
rs1017877420 Chr.8: 63086010-63086010 on Build GRCh38
rs968707940 Chr.8: 63085737-63085737 on Build GRCh38
rs751638180 Chr.8: 63085947-63085947 on Build GRCh38
rs894991464 Chr.8: 63085948-63085948 on Build GRCh38
rs551630439 Chr.8: 63085704-63085704 on Build GRCh38
rs199636231 Chr.8: 63085844-63085844 on Build GRCh38
rs1053935655 Chr.8: 63085691-63085691 on Build GRCh38
rs928983374 Chr.8: 63085891-63085891 on Build GRCh38
rs766592241 Chr.8: 63086062-63086062 on Build GRCh38
rs760014795 Chr.8: 63086003-63086003 on Build GRCh38
rs13257608 Chr.8: 63086015-63086015 on Build GRCh38
rs988005513 Chr.8: 63085789-63085789 on Build GRCh38
rs926590177 Chr.8: 63086078-63086078 on Build GRCh38
rs1055068260 Chr.8: 63086111-63086111 on Build GRCh38
rs750576019 Chr.8: 63085905-63085905 on Build GRCh38
rs1003462662 Chr.8: 63085929-63085929 on Build GRCh38
rs786204758 Chr.8: 63086020-63086020 on Build GRCh38
rs776474801 Chr.8: 63086056-63086056 on Build GRCh38
rs574780098 Chr.8: 63085812-63085812 on Build GRCh38
rs752658141 Chr.8: 63085884-63085884 on Build GRCh38
rs557966488 Chr.8: 63085918-63085918 on Build GRCh38
rs772954067 Chr.8: 63085792-63085792 on Build GRCh38
rs543916738 Chr.8: 63086070-63086070 on Build GRCh38
rs72659829 Chr.8: 63085672-63085672 on Build GRCh38
rs748706307 Chr.8: 63085764-63085764 on Build GRCh38
rs899145715 Chr.8: 63086025-63086025 on Build GRCh38
rs981078515 Chr.8: 63086071-63086071 on Build GRCh38
rs926300177 Chr.8: 63085928-63085928 on Build GRCh38
rs982578184 Chr.8: 63086064-63086064 on Build GRCh38
rs1043210168 Chr.8: 63086085-63086085 on Build GRCh38
rs1013393400 Chr.8: 63085961-63085961 on Build GRCh38
rs566654615 Chr.8: 63085721-63085721 on Build GRCh38
rs535442955 Chr.8: 63085829-63085829 on Build GRCh38
rs770230201 Chr.8: 63085962-63085962 on Build GRCh38
rs994880926 Chr.8: 63086046-63086046 on Build GRCh38
rs760412675 Chr.8: 63085885-63085885 on Build GRCh38
rs977630897 Chr.8: 63085755-63085755 on Build GRCh38
rs920375591 Chr.8: 63086079-63086079 on Build GRCh38
rs562483069 Chr.8: 63086080-63086080 on Build GRCh38
rs533094043 Chr.8: 63086090-63086090 on Build GRCh38

View Full Product Details