Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218275854-218276286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
UniGene
Hs.98475
Gene Chromosome Location
Chr.2: 218270392-218346793 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

Gene Symbol
TMBIM1
Entrez Gene ID
64114
Gene Name
transmembrane BAX inhibitor motif containing 1
Gene Aliases
LFG3, MST100, MSTP100, PP1201, RECS1
UniGene
Hs.591605
Gene Chromosome Location
Chr.2: 218274192-218292577 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

SNP Detail

SNP ID Chromosome Location
rs576676284 Chr.2: 218275982-218275982 on Build GRCh38
rs775743400 Chr.2: 218276068-218276068 on Build GRCh38
rs564115462 Chr.2: 218276005-218276005 on Build GRCh38
rs763029300 Chr.2: 218276078-218276078 on Build GRCh38
rs372231919 Chr.2: 218276251-218276251 on Build GRCh38
rs931590781 Chr.2: 218276221-218276221 on Build GRCh38
rs545805557 Chr.2: 218275992-218275992 on Build GRCh38
rs201500641 Chr.2: 218276030-218276030 on Build GRCh38
rs377493085 Chr.2: 218276009-218276009 on Build GRCh38
rs373122426 Chr.2: 218276153-218276153 on Build GRCh38
rs964159281 Chr.2: 218275914-218275914 on Build GRCh38
rs982845677 Chr.2: 218275995-218275995 on Build GRCh38
rs928249235 Chr.2: 218276001-218276001 on Build GRCh38
rs778277416 Chr.2: 218276017-218276017 on Build GRCh38
rs773813735 Chr.2: 218275951-218275951 on Build GRCh38
rs753868467 Chr.2: 218275992-218275999 on Build GRCh38
rs751664002 Chr.2: 218276125-218276132 on Build GRCh38
rs780220708 Chr.2: 218276107-218276107 on Build GRCh38
rs750337213 Chr.2: 218275999-218275999 on Build GRCh38
rs984420365 Chr.2: 218276224-218276224 on Build GRCh38
rs764082930 Chr.2: 218276086-218276086 on Build GRCh38
rs1027139667 Chr.2: 218275926-218275926 on Build GRCh38
rs146808745 Chr.2: 218276244-218276244 on Build GRCh38
rs759934011 Chr.2: 218276047-218276047 on Build GRCh38
rs1003172831 Chr.2: 218275967-218275967 on Build GRCh38
rs891091976 Chr.2: 218276154-218276154 on Build GRCh38
rs974282773 Chr.2: 218275920-218275920 on Build GRCh38
rs766464557 Chr.2: 218276122-218276122 on Build GRCh38
rs755901521 Chr.2: 218276000-218276000 on Build GRCh38
rs371215328 Chr.2: 218276108-218276108 on Build GRCh38
rs935468182 Chr.2: 218276141-218276141 on Build GRCh38
rs1042924653 Chr.2: 218276110-218276110 on Build GRCh38
rs143728280 Chr.2: 218276144-218276144 on Build GRCh38
rs755138067 Chr.2: 218276124-218276125 on Build GRCh38
rs951188773 Chr.2: 218275969-218275969 on Build GRCh38
rs1034254254 Chr.2: 218275968-218275968 on Build GRCh38
rs1027186687 Chr.2: 218276255-218276255 on Build GRCh38
rs947191311 Chr.2: 218276085-218276085 on Build GRCh38
rs13396404 Chr.2: 218275900-218275900 on Build GRCh38
rs187136282 Chr.2: 218276219-218276219 on Build GRCh38
rs771496510 Chr.2: 218276014-218276014 on Build GRCh38
rs776955799 Chr.2: 218276043-218276043 on Build GRCh38
rs769995398 Chr.2: 218276062-218276062 on Build GRCh38
rs752741794 Chr.2: 218275975-218275975 on Build GRCh38
rs750328598 Chr.2: 218276094-218276094 on Build GRCh38
rs115102659 Chr.2: 218276257-218276257 on Build GRCh38
rs908877590 Chr.2: 218276245-218276246 on Build GRCh38
rs1008195193 Chr.2: 218276187-218276187 on Build GRCh38
rs758382855 Chr.2: 218276023-218276024 on Build GRCh38
rs763007647 Chr.2: 218275981-218275981 on Build GRCh38
rs191778483 Chr.2: 218276253-218276253 on Build GRCh38
rs767115280 Chr.2: 218276179-218276179 on Build GRCh38
rs976008929 Chr.2: 218276188-218276188 on Build GRCh38
rs1021590476 Chr.2: 218276101-218276101 on Build GRCh38
rs1039907947 Chr.2: 218276245-218276245 on Build GRCh38
rs200815904 Chr.2: 218276012-218276012 on Build GRCh38
rs747682844 Chr.2: 218276013-218276013 on Build GRCh38
rs762117520 Chr.2: 218275882-218275882 on Build GRCh38

View Full Product Details