Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113394816-113395089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
UniGene
Hs.448473
Gene Chromosome Location
Chr.11: 113386014-113400418 on Build GRCh38
Transcripts
NM_178510.1 XM_011542736.2 XM_011542737.2 XM_011542738.2 XM_017017475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040617 c.545C>T p.S182L Chr.11: 113394993-113394993 on Build GRCh38
5371671 c.537C>T p.I179I Chr.11: 113394985-113394985 on Build GRCh38
5417086 c.497T>G p.L166R Chr.11: 113394945-113394945 on Build GRCh38
1732879 c.630C>G p.Y210* Chr.11: 113395078-113395078 on Build GRCh38
3985933 c.535A>C p.I179L Chr.11: 113394983-113394983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767086566 Chr.11: 113395000-113395000 on Build GRCh38
rs117599758 Chr.11: 113394873-113394873 on Build GRCh38
rs150678579 Chr.11: 113394986-113394986 on Build GRCh38
rs759009323 Chr.11: 113394998-113394998 on Build GRCh38
rs777568475 Chr.11: 113394959-113394959 on Build GRCh38
rs762261977 Chr.11: 113394993-113394993 on Build GRCh38
rs776054755 Chr.11: 113394899-113394899 on Build GRCh38
rs780291642 Chr.11: 113395042-113395042 on Build GRCh38
rs774031844 Chr.11: 113394995-113394995 on Build GRCh38
rs1027430198 Chr.11: 113394868-113394868 on Build GRCh38
rs375367141 Chr.11: 113394972-113394972 on Build GRCh38
rs373295480 Chr.11: 113394971-113394971 on Build GRCh38
rs763956667 Chr.11: 113395006-113395006 on Build GRCh38
rs769352838 Chr.11: 113395051-113395051 on Build GRCh38
rs751094655 Chr.11: 113394940-113394940 on Build GRCh38
rs769225984 Chr.11: 113394990-113394990 on Build GRCh38
rs756006435 Chr.11: 113394958-113394958 on Build GRCh38
rs772173553 Chr.11: 113394976-113394976 on Build GRCh38
rs749026743 Chr.11: 113394966-113394966 on Build GRCh38
rs754072257 Chr.11: 113394939-113394939 on Build GRCh38
rs752543600 Chr.11: 113394969-113394969 on Build GRCh38
rs550568398 Chr.11: 113395030-113395030 on Build GRCh38
rs561588687 Chr.11: 113395022-113395022 on Build GRCh38
rs779011394 Chr.11: 113395001-113395001 on Build GRCh38
rs777083250 Chr.11: 113394924-113394924 on Build GRCh38
rs753702547 Chr.11: 113395014-113395014 on Build GRCh38
rs371127439 Chr.11: 113395025-113395025 on Build GRCh38
rs369211659 Chr.11: 113394905-113394905 on Build GRCh38
rs577943707 Chr.11: 113394941-113394941 on Build GRCh38
rs764581391 Chr.11: 113394907-113394907 on Build GRCh38
rs897218854 Chr.11: 113395010-113395010 on Build GRCh38
rs775667496 Chr.11: 113394985-113394985 on Build GRCh38
rs949855351 Chr.11: 113395012-113395012 on Build GRCh38
rs777136605 Chr.11: 113394991-113394991 on Build GRCh38
rs868573335 Chr.11: 113394896-113394896 on Build GRCh38
rs747016489 Chr.11: 113395043-113395043 on Build GRCh38
rs140803658 Chr.11: 113394994-113394994 on Build GRCh38
rs115800217 Chr.11: 113395002-113395002 on Build GRCh38
rs994154547 Chr.11: 113394855-113394855 on Build GRCh38
rs757065955 Chr.11: 113395021-113395021 on Build GRCh38
rs760473928 Chr.11: 113395027-113395027 on Build GRCh38
rs758884619 Chr.11: 113395037-113395037 on Build GRCh38
rs920474257 Chr.11: 113394982-113394982 on Build GRCh38
rs767292061 Chr.11: 113394918-113394918 on Build GRCh38
rs375895682 Chr.11: 113394898-113394898 on Build GRCh38
rs767343068 Chr.11: 113394927-113394927 on Build GRCh38
rs752600844 Chr.11: 113394956-113394956 on Build GRCh38
rs961779358 Chr.11: 113394846-113394846 on Build GRCh38
rs1032517468 Chr.11: 113394872-113394872 on Build GRCh38
rs200549519 Chr.11: 113394925-113394925 on Build GRCh38
rs865792934 Chr.11: 113395036-113395036 on Build GRCh38
rs141919233 Chr.11: 113394945-113394945 on Build GRCh38
rs1019657342 Chr.11: 113394841-113394841 on Build GRCh38
rs745998002 Chr.11: 113394974-113394974 on Build GRCh38
rs745898839 Chr.11: 113394885-113394885 on Build GRCh38

View Full Product Details