Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26464051-26464302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
UniGene
Hs.474436
Gene Chromosome Location
Chr.22: 26443613-26483863 on Build GRCh38
Transcripts
NM_022081.5 NM_152841.2 NR_073135.1 NR_073136.1 XM_006724353.3 XM_006724354.3 XM_006724360.3 XM_011530485.1 XM_011530486.2 XM_011530487.2 XM_011530488.2 XM_011530489.2 XM_011530490.2 XM_011530491.2 XM_011530492.1 XM_011530493.2 XM_011530494.2 XM_011530495.2 XM_011530496.2 XM_017029042.1 XM_017029043.1 XM_017029044.1 XM_017029045.1 XM_017029046.1 XM_017029047.1 XM_017029048.1 XM_017029049.1 XM_017029050.1 XM_017029051.1 XM_017029052.1 XM_017029053.1 XM_017029054.1 XM_017029055.1 XM_017029056.1 XM_017029057.1 XM_017029058.1 XM_017029059.1 XM_017029060.1 XM_017029061.1 XM_017029062.1 XM_017029063.1 XM_017029064.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2935537 c.1466C>T p.P489L Chr.22: 26464203-26464203 on Build GRCh38
4691091 c.1539C>T p.S513S Chr.22: 26464130-26464130 on Build GRCh38
5651995 c.1519G>C p.E507Q Chr.22: 26464111-26464111 on Build GRCh38
6681973 c.1525G>A p.G509R Chr.22: 26464144-26464144 on Build GRCh38
6526433 c.1458G>T p.G486G Chr.22: 26464172-26464172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772234990 Chr.22: 26464176-26464176 on Build GRCh38
rs868526867 Chr.22: 26464241-26464241 on Build GRCh38
rs375587915 Chr.22: 26464260-26464260 on Build GRCh38
rs763311479 Chr.22: 26464110-26464110 on Build GRCh38
rs988050148 Chr.22: 26464178-26464178 on Build GRCh38
rs767852159 Chr.22: 26464126-26464126 on Build GRCh38
rs549290787 Chr.22: 26464219-26464219 on Build GRCh38
rs774532334 Chr.22: 26464119-26464119 on Build GRCh38
rs186173240 Chr.22: 26464270-26464270 on Build GRCh38
rs372833027 Chr.22: 26464084-26464084 on Build GRCh38
rs141008676 Chr.22: 26464094-26464094 on Build GRCh38
rs772205948 Chr.22: 26464247-26464247 on Build GRCh38
rs148134252 Chr.22: 26464087-26464087 on Build GRCh38
rs367810755 Chr.22: 26464097-26464097 on Build GRCh38
rs142958812 Chr.22: 26464233-26464233 on Build GRCh38
rs770822747 Chr.22: 26464103-26464103 on Build GRCh38
rs752092814 Chr.22: 26464148-26464148 on Build GRCh38
rs965241113 Chr.22: 26464222-26464222 on Build GRCh38
rs893836600 Chr.22: 26464125-26464125 on Build GRCh38
rs756030696 Chr.22: 26464236-26464236 on Build GRCh38
rs147435410 Chr.22: 26464234-26464234 on Build GRCh38
rs768468362 Chr.22: 26464277-26464279 on Build GRCh38
rs983056340 Chr.22: 26464254-26464254 on Build GRCh38
rs11545917 Chr.22: 26464143-26464143 on Build GRCh38
rs757988379 Chr.22: 26464150-26464150 on Build GRCh38
rs957749322 Chr.22: 26464269-26464269 on Build GRCh38
rs754462097 Chr.22: 26464091-26464091 on Build GRCh38
rs761197874 Chr.22: 26464187-26464187 on Build GRCh38
rs777570870 Chr.22: 26464151-26464151 on Build GRCh38
rs778544549 Chr.22: 26464142-26464142 on Build GRCh38
rs748136926 Chr.22: 26464274-26464274 on Build GRCh38
rs769017976 Chr.22: 26464116-26464116 on Build GRCh38
rs754224646 Chr.22: 26464137-26464137 on Build GRCh38
rs773570398 Chr.22: 26464180-26464180 on Build GRCh38
rs768100817 Chr.22: 26464073-26464073 on Build GRCh38
rs756215705 Chr.22: 26464175-26464175 on Build GRCh38
rs764860798 Chr.22: 26464216-26464216 on Build GRCh38
rs756512740 Chr.22: 26464076-26464076 on Build GRCh38
rs1034179775 Chr.22: 26464120-26464120 on Build GRCh38
rs773303744 Chr.22: 26464252-26464252 on Build GRCh38
rs774728501 Chr.22: 26464166-26464166 on Build GRCh38
rs1021234084 Chr.22: 26464164-26464164 on Build GRCh38
rs754066787 Chr.22: 26464245-26464245 on Build GRCh38
rs745687268 Chr.22: 26464163-26464163 on Build GRCh38
rs139039617 Chr.22: 26464212-26464212 on Build GRCh38
rs372027593 Chr.22: 26464235-26464235 on Build GRCh38
rs757812079 Chr.22: 26464227-26464227 on Build GRCh38
rs763704933 Chr.22: 26464228-26464228 on Build GRCh38
rs775820827 Chr.22: 26464106-26464106 on Build GRCh38
rs751051493 Chr.22: 26464231-26464231 on Build GRCh38
rs766781092 Chr.22: 26464128-26464128 on Build GRCh38
rs761956374 Chr.22: 26464121-26464121 on Build GRCh38
rs781082499 Chr.22: 26464158-26464158 on Build GRCh38
rs778346173 Chr.22: 26464092-26464092 on Build GRCh38
rs534067573 Chr.22: 26464127-26464127 on Build GRCh38
rs768931718 Chr.22: 26464165-26464165 on Build GRCh38
rs770823084 Chr.22: 26464152-26464152 on Build GRCh38
rs766903925 Chr.22: 26464188-26464188 on Build GRCh38
rs942076085 Chr.22: 26464173-26464173 on Build GRCh38
rs759909785 Chr.22: 26464213-26464213 on Build GRCh38
rs778998363 Chr.22: 26464243-26464243 on Build GRCh38
rs150216540 Chr.22: 26464095-26464095 on Build GRCh38
rs750685060 Chr.22: 26464074-26464074 on Build GRCh38

View Full Product Details