Gene Transcripts

Gene Symbol
Grhl3
Entrez Gene ID
230824
Gene Name
grainyhead-like 3 (Drosophila)
Gene Aliases
AI561912, Get1, Som, Tfcp2l4, ct
Chromosome Location
Chr. 4: 135541887 - 135573628 on Build GRCm38
UniGene ID
Mm.331972
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013756.1 NP_001013778.1 4 545
GenBank mRNA BC089372.1 - 4 545

View Full Product Details