Gene Transcripts

Gene Symbol
Mapk9
Entrez Gene ID
26420
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
AI851083, JNK2, Prkm9, p54aSAPK
Chromosome Location
Chr. 11: 49846751 - 49886421 on Build GRCm38
UniGene ID
Mm.68933
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163671.1 NP_001157143.1 6 800
NM_001163672.1 NP_001157144.1 6 800
NM_016961.3 NP_058657.1 6 800
NM_207692.2 NP_997575.2 6 800
GenBank mRNA AB005664.1 - 5 574
AF052466.1 - 5 536
AF052467.1 - 5 536
AF052468.1 - 5 536
AF052469.1 - 5 536
AK031959.1 - 6 799
AK031974.1 - 4 354
AK043169.1 - 6 658
AK090057.1 - 6 744
AK145083.1 - 6 775
BC028341.1 - 6 782 4192796

View Full Product Details