Gene Transcripts

Gene Symbol
Flrt1
Entrez Gene ID
396184
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
AW742165, D630040I23Rik
Chromosome Location
Chr. 19: 7087464 - 7166030 on Build GRCm38
UniGene ID
Mm.386930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201411.2 NP_958813.1 2 2055
GenBank mRNA AK031503.1 - 2 2055
AY495667.1 - 1 141
AY495668.1 - 2 2010
BC070403.1 - 2 2059 5696478
BC111879.1 - 2 1758 40059728
BC112383.1 - 2 1758 40059727
BC138214.1 - 2 1696 8861233

View Full Product Details