Gene Transcripts

Gene Symbol
Klhdc9
Entrez Gene ID
68874
Gene Name
kelch domain containing 9
Gene Aliases
1190002J23Rik, AA087357, ESTM31
Chromosome Location
Chr. 1: 171358449 - 171360798 on Build GRCm38
UniGene ID
Mm.10756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033039.2 NP_001028211.1 3 945
GenBank mRNA AK076812.1 - 3 668
AK140290.1 - 3 946
BC141196.1 - 3 858 9055808

View Full Product Details