Gene Transcripts

Gene Symbol
Jmjd8
Entrez Gene ID
72106
Gene Name
jumonji domain containing 8
Gene Aliases
2610003J06Rik
Chromosome Location
Chr. 17: 25829043 - 25831843 on Build GRCm38
UniGene ID
Mm.277599
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028101.4 NP_082377.2 7 599
GenBank mRNA AK011308.1 - 7 596
AK148079.1 - 7 577
AK155810.1 - 7 601
AK172073.1 - 7 596
BC010800.1 - 7 561 4235072

View Full Product Details