Gene Transcripts

Gene Symbol
Tsen54
Entrez Gene ID
76265
Gene Name
tRNA splicing endonuclease 54 homolog (S. cerevisiae)
Gene Aliases
0610034P02Rik
Chromosome Location
Chr. 11: 115813743 - 115823102 on Build GRCm38
UniGene ID
Mm.275438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029557.1 NP_083833.1 7 578
GenBank mRNA AK002758.1 - 7 563
AK089005.1 - 7 545
BC126953.1 - 6 702

View Full Product Details