Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024471.4 | NP_077791.1 | 8 | 940 | |
GenBank mRNA | AB057731.1 | - | 8 | 839 | |
AK031851.1 | - | 8 | 912 | ||
AK045167.1 | - | 8 | 725 | ||
AK048744.1 | - | 884 | |||
AK049548.1 | - | 8 | 855 | ||
AK077192.1 | - | 8 | 941 | ||
BC002141.1 | - | 8 | 872 | 3484559 |