Gene Transcripts

Gene Symbol
Prss28
Entrez Gene ID
114661
Gene Name
protease, serine 28
Gene Aliases
Isp1, mIsp-1
Chromosome Location
Chr. 17: 25308646 - 25311876 on Build GRCm38
UniGene ID
Mm.207081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053259.2 NP_444489.2 5 702
GenBank mRNA AF184895.1 - 4 541
AK144498.1 - 5 703
BC119386.1 - 4 569 8734135
BC119388.1 - 4 569 8734137

View Full Product Details