Gene Transcripts

Gene Symbol
Adck5
Entrez Gene ID
268822
Gene Name
aarF domain containing kinase 5
Gene Aliases
A230108P17
Chromosome Location
Chr. 15: 76576359 - 76595812 on Build GRCm38
UniGene ID
Mm.181624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172960.3 NP_766548.2 15 1857
GenBank mRNA AK039217.1 - 15 1853
AK163557.1 - 14 1884
BC030881.1 - 12 2080
BC053696.1 - 5 1045
BC060528.1 - 15 1741 6414642

View Full Product Details