Gene Transcripts

Gene Symbol
Trrap
Entrez Gene ID
100683
Gene Name
transformation/transcription domain-associated protein
Gene Aliases
AI481500
Chromosome Location
Chr. 5: 144768097 - 144859778 on Build GRCm38
UniGene ID
Mm.273570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081362.1 NP_001074831.1 58, 59 9067
GenBank mRNA AK147241.1 - 58, 59 9067
AK147613.1 - 35, 36 5583
BC029023.1 - 11, 12 1814

View Full Product Details