Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008231.4 | NP_032257.3 | 3 | 580 | |
GenBank mRNA | AF251787.1 | - | 3 | 493 | |
AK017863.1 | - | 3 | 534 | ||
AK029475.1 | - | 3 | 324 | ||
AK076021.1 | - | 3 | 368 | ||
AK185514.1 | - | 2 | 192 | ||
BC005713.1 | - | 3 | 490 | 2609261 | |
BC021654.1 | - | 3 | 508 | 4163897 | |
D63707.1 | - | 3 | 334 |