Gene Transcripts
- Gene Symbol
- Urb1
- Entrez Gene ID
- 207932
- Gene Name
- URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
- Gene Aliases
- 4921511H13Rik, 5730405K23Rik, AA670781, AI645590, mKIAA0539
- Chromosome Location
- Chr. 16: 90751527 - 90810465 on Build GRCm38
- UniGene ID
- Mm.328688
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029497.1 |
NP_083773.1
|
12 |
1694 |
|
GenBank mRNA |
AK050735.1 |
- |
12 |
1674 |
|
View Full Product Details