Gene Transcripts
- Gene Symbol
- Timp1
- Entrez Gene ID
- 21857
- Gene Name
- tissue inhibitor of metalloproteinase 1
- Gene Aliases
- Clgi, EPA, TIMP-1, TPA-S1, Timp
- Chromosome Location
- Chr. X: 20870166 - 20874737 on Build GRCm38
- UniGene ID
- Mm.8245
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001044384.1 |
NP_001037849.1
|
5, 6 |
624 |
|
NM_001294280.2 |
NP_001281209.1
|
4, 5 |
544 |
|
NM_011593.2 |
NP_035723.2
|
5, 6 |
621 |
|
GenBank mRNA |
AA184230.1 |
- |
3, 4 |
390 |
|
AY622853.1 |
- |
2, 3 |
158 |
|
BC008107.1 |
- |
5, 6 |
526 |
3158222 |
BC034260.1 |
- |
5, 6 |
526 |
3158222 |
BC051260.1 |
- |
5, 6 |
506 |
30056565 |
BU756519.1 |
- |
1, 2 |
278 |
|
M17243.1 |
- |
5, 6 |
487 |
|
X04684.1 |
- |
5, 6 |
563 |
|
View Full Product Details