Gene Transcripts

Gene Symbol
Vegfb
Entrez Gene ID
22340
Gene Name
vascular endothelial growth factor B
Gene Aliases
VEGF-B, Vrf
Chromosome Location
Chr. 19: 6982472 - 6987651 on Build GRCm38
UniGene ID
Mm.15607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185164.1 NP_001172093.1 3 491
NM_011697.3 NP_035827.1 3 491
GenBank mRNA AK148188.1 - 3 490
AK182808.1 - 1 120
AK207652.1 - 1 118
AK208173.1 - 1 122
AK211743.1 - 1 123
AK212120.1 - 1 122
AK216634.1 - 1 122
BC046303.1 - 3 469 6492277
U43836.1 - 3 443
U43837.1 - 3 251
U48800.1 - 3 345
U52820.1 - 3 345

View Full Product Details