Gene Transcripts

Gene Symbol
Agfg2
Entrez Gene ID
231801
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
A630095P14Rik, Hrbl, RAB-R, RABR
Chromosome Location
Chr. 5: 137650463 - 137684806 on Build GRCm38
UniGene ID
Mm.260869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK155221.1 - 1 287
AK171048.1 - 1 332
BC003330.1 - 1 299 3256586
BC046788.1 - 1 223 5716604

View Full Product Details