Gene Transcripts
- Gene Symbol
- Gcnt7
- Entrez Gene ID
- 654821
- Gene Name
- glucosaminyl (N-acetyl) transferase family member 7
- Gene Aliases
- A330041C17Rik, gnt
- Chromosome Location
- Chr. 2: 172447120 - 172458596 on Build GRCm38
- UniGene ID
- Mm.389337
- Species
- Mus musculus
- Gene Symbol
- Rtfdc1
- Entrez Gene ID
- 66404
- Gene Name
- replication termination factor 2 domain containing 1
- Gene Aliases
- 1700067C04Rik, 2410001C21Rik, 5730427M17Rik, AA589417
- Chromosome Location
- Chr. 2: 172440578 - 172469899 on Build GRCm38
- UniGene ID
- Mm.25157
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030734.1 |
- |
1 |
4062 |
|
View Full Product Details