Gene Transcripts

Gene Symbol
Vom2r73
Entrez Gene ID
689670
Gene Name
vomeronasal 2 receptor, 73
Gene Aliases
Chromosome Location
Chr. 14: 1773425 - 1791636 on Build Rnor_6.0
UniGene ID
Rn.216132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099486.1 NP_001092956.1 3 687

View Full Product Details