Gene Transcripts

Gene Symbol
Pcf11
Entrez Gene ID
361605
Gene Name
PCF11 cleavage and polyadenylation factor subunit
Gene Aliases
RGD1563397
Chromosome Location
Chr. 1: 157463014 - 157494130 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759657.1 XP_008757879.1 2 534
XM_341883.6 XP_341884.4 2 534

View Full Product Details