Gene Transcripts

Gene Symbol
Shb
Entrez Gene ID
362513
Gene Name
Src homology 2 domain containing adaptor protein B
Gene Aliases
RGD1565350
Chromosome Location
Chr. 5: 60970650 - 61077881 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238148.1 XP_006238210.1 5 1499
XM_008763691.1 XP_008761913.1 5 1331

View Full Product Details