Gene Transcripts

Gene Symbol
Aurka
Entrez Gene ID
261730
Gene Name
aurora kinase A
Gene Aliases
ARK-1, Stk6
Chromosome Location
Chr. 3: 170361529 - 170380277 on Build Rnor_6.0
UniGene ID
Rn.161874
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153296.2 NP_695208.2 5, 6 689
XM_006235675.2 XP_006235737.1 6, 7 768
XM_006235676.2 XP_006235738.1 6, 7 785
GenBank mRNA AF537333.1 - 5, 6 677
BC104676.1 - 5, 6 689 7453115
FQ227101.1 - 6, 7 775

View Full Product Details