Gene Transcripts

Gene Symbol
Ptar1
Entrez Gene ID
286972
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
Chromosome Location
Chr. 1: 241501679 - 241557651 on Build Rnor_6.0
UniGene ID
Rn.107234
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105760.2 NP_001099230.2 5 583

View Full Product Details