Gene Transcripts

Gene Symbol
Trim52
Entrez Gene ID
290458
Gene Name
tripartite motif-containing 52
Gene Aliases
RGD1565615
Chromosome Location
Chr. 15: 90364514 - 90387915 on Build Rnor_6.0
UniGene ID
Rn.218506
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106056.1 NP_001099526.1 3 130
XM_006252435.2 XP_006252497.1 1 1021

View Full Product Details