Gene Transcripts

Gene Symbol
Rtdr1
Entrez Gene ID
365552
Gene Name
rhabdoid tumor deletion region gene 1
Gene Aliases
Chromosome Location
Chr. 20: 14577122 - 14653057 on Build Rnor_6.0
UniGene ID
Rn.123513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127557.2 NP_001121029.1 1 64
XM_006256340.2 XP_006256402.1 2 120
XM_006256341.2 XP_006256403.1 2 173
GenBank mRNA BC162057.1 - 1 66

View Full Product Details