Gene Transcripts

Gene Symbol
Nufip2
Entrez Gene ID
681371
Gene Name
nuclear fragile X mental retardation protein interacting protein 2
Gene Aliases
Chromosome Location
Chr. 10: 65175344 - 65200215 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750867.3 XP_003750915.1 2 1662

View Full Product Details