Gene Transcripts
- Gene Symbol
- MTHFD2
- Entrez Gene ID
- 10797
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase
- Gene Aliases
- NMDMC
- Chromosome Location
- Chr. 2: 74198563 - 74215298 on Build GRCh38
- UniGene ID
- Hs.469030
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027405.1 |
|
2, 3 |
275 |
|
GenBank mRNA |
BC015062.1 |
- |
2, 3 |
225 |
|
View Full Product Details