Gene Transcripts
- Gene Symbol
- PDS5A
- Entrez Gene ID
- 23244
- Gene Name
- PDS5 cohesin associated factor A
- Gene Aliases
- PIG54, SCC-112, SCC112
- Chromosome Location
- Chr. 4: 39822863 - 39977956 on Build GRCh38
- UniGene ID
- Hs.331431
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124569.1 |
- |
1 |
426 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC131566.1 |
- |
1 |
61 |
|
Chr. 3 - 63925120 - 63925632 [-] |
View Full Product Details