Gene Transcripts
- Gene Symbol
- THNSL2
- Entrez Gene ID
- 55258
- Gene Name
- threonine synthase like 2
- Gene Aliases
- SOFAT, THS2, TSH2
- Chromosome Location
- Chr. 2: 88170295 - 88186637 on Build GRCh38
- UniGene ID
- Hs.739080
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092823.1 |
- |
1 |
1091 |
|
View Full Product Details