Gene Transcripts

Gene Symbol
MAGI2-AS3
Entrez Gene ID
100505881
Gene Name
MAGI2 antisense RNA 3
Gene Aliases
Chromosome Location
Chr. 7: 79452957 - 79471208 on Build GRCh38
UniGene ID
Hs.31474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038343.2 4 2826
GenBank mRNA AK021837.1 - 1 290
AL833456.1 - 2 2551
BX641113.1 - 1 1718
CR749529.1 - 2 2879

View Full Product Details