Gene Transcripts

Gene Symbol
NKD1
Entrez Gene ID
85407
Gene Name
naked cuticle homolog 1
Gene Aliases
Naked1
Chromosome Location
Chr. 16: 50548330 - 50649249 on Build GRCh38
UniGene ID
Hs.187578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033119.4 NP_149110.1 10 6573
XM_011523399.2 XP_011521701.1 13 6884
XM_011523400.2 XP_011521702.1 8 6175
XM_017023804.1 XP_016879293.1 12 8103
GenBank mRNA AK021701.1 - 1 850
AK023784.1 - 1 1125

View Full Product Details