Gene Transcripts
- Gene Symbol
- Msl3l2
- Entrez Gene ID
- 73390
- Gene Name
- male-specific lethal 3-like 2 (Drosophila)
- Gene Aliases
- 1700060H10Rik
- Chromosome Location
- Chr. 10: 56083827 - 56116880 on Build GRCm38
- UniGene ID
- Mm.181475
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK172141.1 |
- |
|
743 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY765891.1 |
- |
1 |
235 |
|
Chr. 10 - 56116866 - 56117573 [+] |
View Full Product Details