Gene Transcripts

Gene Symbol
Epb4.1
Entrez Gene ID
269587
Gene Name
erythrocyte protein band 4.1
Gene Aliases
4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056
Chromosome Location
Chr. 4: 131923413 - 132075579 on Build GRCm38
UniGene ID
Mm.30038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK148569.1 - 1 2176


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ042489.1 - 1 417 Chr. 4 - 131934278 - 131935018 [-]

View Full Product Details