Gene Transcripts
- Gene Symbol
- Erbb2
- Entrez Gene ID
- 13866
- Gene Name
- v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
- Gene Aliases
- Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023
- Chromosome Location
- Chr. 11: 98412484 - 98437716 on Build GRCm38
- UniGene ID
- Mm.290822
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK160975.1 |
- |
|
2002 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW211491.1 |
- |
1 |
248 |
|
Chr. 11 - 98438927 - 98439410 [+] |
GenBankmRNA |
BI647657.1 |
- |
1 |
586 |
|
Chr. 11 - 98438595 - 98439384 [+] |
GenBankmRNA |
BI650383.1 |
- |
|
268 |
|
Chr. - - [-] |
GenBankmRNA |
AK027956.1 |
- |
|
603 |
|
Chr. - - [-] |
GenBankmRNA |
BI645894.1 |
- |
1 |
595 |
|
Chr. 11 - 98438580 - 98439476 [+] |
GenBankmRNA |
BI108279.1 |
- |
|
269 |
|
Chr. - - [-] |
View Full Product Details