Gene Transcripts

Gene Symbol
Adamts20
Entrez Gene ID
223838
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 20
Gene Aliases
Adamst20, bt
Chromosome Location
Chr. 15: 94268314 - 94465427 on Build GRCm38
UniGene ID
Mm.214132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164785.1 NP_001158257.1 26 3986
NM_177431.4 NP_803180.3 26 3986
GenBank mRNA AJ512753.3 - 26 3755
AK142029.1 - 7 904
AY189815.1 - 26 3894
AY189816.1 - 26 3894
BC121792.1 - 26 3825 40099393

View Full Product Details