Gene Transcripts

Gene Symbol
Usp40
Entrez Gene ID
227334
Gene Name
ubiquitin specific peptidase 40
Gene Aliases
B230215L03Rik, C730029K03
Chromosome Location
Chr. 1: 87945121 - 88008577 on Build GRCm38
UniGene ID
Mm.80484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033291.2 NP_001028463.1 7 921
NM_001198573.1 NP_001185502.1 7 921
GenBank mRNA AK050239.1 - 7 918
AK132295.1 - 7 921
AK166303.1 - 7 898
BC030843.1 - 7 870
BC089376.1 - 4 423
BC117957.1 - 8 1036 40098174
BC117958.1 - 7 904 40098175

View Full Product Details