Gene Transcripts

Gene Symbol
Ffar3
Entrez Gene ID
233080
Gene Name
free fatty acid receptor 3
Gene Aliases
Gm478, Gpr41
Chromosome Location
Chr. 7: 30854330 - 30856178 on Build GRCm38
UniGene ID
Mm.291167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033316.2 NP_001028488.1 2 978
GenBank mRNA AK148616.1 - 2 979
BC109330.2 - 1 1191 40051470
BC125009.1 - 1 1191 40051462

View Full Product Details