Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164272.1 | NP_001157744.1 | 4 | 455 | |
NM_172302.3 | NP_758506.3 | 4 | 455 | ||
GenBank mRNA | AK044306.1 | - | 4 | 435 | |
AK053375.1 | - | 5 | 500 | ||
AK088603.1 | - | 4 | 433 | ||
AK165330.1 | - | 4 | 433 | ||
AK184839.1 | - | 2 | 85 | ||
BC017519.1 | - | 3 | 375 | ||
BC038812.1 | - | 3 | 385 | 4506179 |