Gene Transcripts

Gene Symbol
PVALB
Entrez Gene ID
5816
Gene Name
parvalbumin
Gene Aliases
D22S749
Chromosome Location
Chr. 22: 36800701 - 36819473 on Build GRCh38
UniGene ID
Hs.295449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001315532.1 NP_001302461.1 2 154
NM_002854.2 NP_002845.1 3 174
GenBank mRNA AK311832.1 - 2 154
AV724364.1 - 2 133
BC069300.1 - 3 174 7262744
BC096112.3 - 3 180 40002305
BC096113.3 - 3 180 40002307
BC096114.1 - 3 180 40002308
BC096115.1 - 3 180 40002310
CR456552.1 - 2 140
DA392417.1 - 2 154
X63070.1 - 3 180

View Full Product Details