Gene Transcripts

Gene Symbol
Gal3st4
Entrez Gene ID
330217
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
1500031A01Rik
Chromosome Location
Chr. 5: 138264918 - 138273038 on Build GRCm38
UniGene ID
Mm.332774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033416.2 NP_001028588.1 3 2712
GenBank mRNA AK132558.1 - 3 2713
AK138993.1 - 4 1377
AK162468.1 - 1 937

View Full Product Details