Gene Transcripts

Gene Symbol
Aqp4
Entrez Gene ID
25293
Gene Name
aquaporin 4
Gene Aliases
AQP-4, MIWC2, Miwc
Chromosome Location
Chr. 18: 6766009 - 6782757 on Build Rnor_6.0
UniGene ID
Rn.90091
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142366.2 NP_001135838.1 4 718
NM_001270558.2 NP_001257487.1 3 545
NM_001270559.2 NP_001257488.1 3 553
NM_012825.4 NP_036957.1 4 710
GenBank mRNA L27588.1 - 3 578
U14007.1 - 4 644

View Full Product Details